Ac-YVAD-CHO
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260027M001 ALX260027M005
ENZOALX260027M001EA 90.2 EUR
ENZOALX260027M001 ENZOALX260027M005
Ac-YVAD-CHO
Eiwitten en peptiden
Inhibitor of caspase-1 and -4
Potent and reversible inhibitor of caspase-1. Also inhibits caspase-4. Strongly inhibits anti-Fas induced apoptosis in L929-Fas cells.
Order Now
Specificaties
Protein/peptide name Ac-YVAD-CHO Ac-YVAD-CHO
Environmentally Preferable
Species
Bron
Zuiverheid >96% (HPLC) >96% (HPLC)
Formulering Lyophilized __
Reconstitution instructions Add 0,2 ml DMSO per 1,0 mg peptide. Shake well until all the contents are dissolved. This furnishes a 10 mM solution of this material. __