Ac-WEHD-CHO
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260055M001 ALX260055M005
ENZOALX260055M001EA 174 EUR
ENZOALX260055M001 ENZOALX260055M005
Ac-WEHD-CHO
Eiwitten en peptiden
Inhibitor of caspase-1, -4 and -5
Reversible, cell permeable inhibitor of caspase-1 and -5. Also inhibits caspase-4.