N-(1-Naphthalenylsulfonyl)-Ile-Trp-aldehyde
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260133M001 ALX260133M005
ENZOALX260133M001EA 209 EUR
ENZOALX260133M001 ENZOALX260133M005
N-(1-Naphthalenylsulfonyl)-Ile-Trp-aldehyde
Eiwitten en peptiden
Cathepsin-L inhibitor
Potent, selective and reversible inhibitor of human cathepsin L (IC50=1,9 nM). Inhibits release of Ca²⁺ and hydroxyproline from bone in vitro.