Z-VAD-FMK
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260138R020 ALX260138R100
ENZOALX260138R020EA 281 EUR
ENZOALX260138R020 ENZOALX260138R100
Z-VAD-FMK
Eiwitten en peptiden
Caspase inhibitor
Synthetic peptide that irreversibly inhibits ICE-family protease/caspase activity and blocks apoptosis. The inhibitor is designed as a methyl ester to facilitate cell permeability.