Z-YVAD-FMK
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260154R020 ALX260154R100
ENZOALX260154R020EA 275 EUR
ENZOALX260154R020 ENZOALX260154R100
Z-YVAD-FMK
Eiwitten en peptiden
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4.