Z-VRPR-FMK
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260166M001 ALX260166MC05
ENZOALX260166M001EA 753 EUR
ENZOALX260166M001 ENZOALX260166MC05
Z-VRPR-FMK
Eiwitten en peptiden
MALT1 inhibitor
The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity.