Microcystin-RR
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX350043C050 ALX350043C100 ALX350043C250 ALX350043C500 ALX350043M001
ENZOALX350043C050EA 84.7 EUR
ENZOALX350043C050 ENZOALX350043C100 ENZOALX350043C250 ENZOALX350043C500 ENZOALX350043M001
Microcystin-RR
Eiwitten en peptiden
PP2A inhibitor
Arg-Arg analog of microcystin-LR (Prod. No. ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A).
Isolated from Microcystis aeruginosa.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9f3e327536ee465993549b60863ccc73893029&vid=35c07ea1a8f8a5239f0cae5deda073334c52a55e&prid=web&clid=SAPDAM