Microcystin-YR
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX350044C025 ALX350044C100
ENZOALX350044C025EA 275 EUR
ENZOALX350044C025 ENZOALX350044C100
Microcystin-YR
Eiwitten en peptiden
Inhibitor of PP1 and PP2A
Analog of microcystin-LR (Prod. No. ALX-350-012) with Tyr substituted in place of Leu. As for all microcystins the conjugated double bonds in the ADDA moiety cause a characteristic absorption maximum at 238 nm. The Tyr residue in position 2 of microcystin-YR confers an absorption maximum at 232 nm. Potent inhibitor of eukaryotic protein phosphatases 1 and 2 A. Useful as a reference compound in environmental analysis. The hydroxyl group of the Tyr residue may prove useful for linking microcystin-YR via conjugation to other chemicals. Inhibits the synthesis of proteases such as cathepsin D and L, and arginine aminopeptidase.
Isolated from Microcystis aeruginosa.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbaa3ea7164160bf3626aa0b69690dad224ec6ce&vid=0fca380f5c729632a21ea3b4ceed8403035434d7&prid=web&clid=SAPDAM