Microcystin-LA
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX350096C025 ALX350096C100
ENZOALX350096C025EA 261 EUR
ENZOALX350096C025 ENZOALX350096C100
Microcystin-LA
Eiwitten en peptiden
Inhibitor of PP2A and calcineurin
Analog of microcystin-LR (Prod. No. ALX-350-012) with Ala substituted in place of Arg. Inhibits protein phosphatase 2A (PP2A) and protein phosphatase 3 (PP3) more potently than protein phosphatase 1 (PP1). Ozonation of toxic water samples does lead to a complete loss of the microcystins and their toxicity.
Isolated from Microcystis aeruginosa.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef9d3082ea6c92cabbac44b1d9af720c26cd57b6&vid=a5a5ac66ef6547fc41e0c401711ab63ee23f742a&prid=web&clid=SAPDAM