N-Acetyl-Leu-Leu-Methional
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1000005 BMLPI1000025
ENZOBMLPI1000005EA 165 EUR
ENZOBMLPI1000005 ENZOBMLPI1000025
N-Acetyl-Leu-Leu-Methional
Eiwitten en peptiden
Calpain inhibitor
Cell-permeable, peptide aldehyde inhibitor of calpain and other neutral cysteine proteases. Potent inhibitor of cathepsin L (Ki=0,6 nM) and cathepsin B (Ki=100 nM).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dfca3629b745f282b87ad6444eb2cd2e0cad18f&vid=f91b049eb6b004ba2c5a0a81fa6039fcc5f83b0c&prid=web&clid=SAPDAM