E-64-D
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1070001
BMLPI1070005
ENZOBMLPI1070001EA
86.9
EUR
ENZOBMLPI1070001
ENZOBMLPI1070005
E-64-D
Eiwitten en peptiden
Calpain and cathepsin inhibitor
Cell-permeable, ethyl ester of E-64-c. The ester is hydrolyzed by intracellular esterases to produce E-64-c. Inhibits calpain and cathepsins B, H, and L. Inhibits degradation of autophagic cargo inside autophagolysosomes.