Phosphoramidon
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1110005 BMLPI1110025
ENZOBMLPI1110005EA 211 EUR
ENZOBMLPI1110005 ENZOBMLPI1110025
Phosphoramidon
Eiwitten en peptiden
Neprilysin inhibitor
Inhibits neutral endopeptidase 24,11 (NEP, neprilysin; EC 3.4.24.11) (Ki=2 nM), a cell surface zinc metalloprotease which cleaves bioactive peptides such as bradykinin, neurotensin, and enkephalin. Also inhibits neprilysin 2 (Ki~2 nM). Potent inhibitor of endothelin-converting enzyme (ECE) (IC50=0,68 μM).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7be80807c69036652df1003d117ba57b9660b47d&vid=3755436f598b5097ac22c9b58935871f8d695d53&prid=web&clid=SAPDAM