Boc-Gly-Val-Val-CHO
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1280005 BMLPI1280025
ENZOBMLPI1280005EA 138 EUR
ENZOBMLPI1280005 ENZOBMLPI1280025
Boc-Gly-Val-Val-CHO
Eiwitten en peptiden
gamma-secretase inhibitor
A novel substrate-based (gamma-40 site) aldehyde inhibitor of gamma-secretase. In whole cells overexpressing amloid ß protein precursor, Boc-GVV-CHO dose-dependently inhibited amyloid ß cleavage. gamma-secretase is an important therapeutic target for strategies designed to lower amyloid ß production.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7f3bf5c0e713f3b0e4dad27dd30005a6c161467&vid=4c69dfe8c40f802638b71d9f1207a6c0ac4d7ef9&prid=web&clid=SAPDAM