Boc-Gly-Val-Val-CHO
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1280005
BMLPI1280025
ENZOBMLPI1280005EA
138
EUR
ENZOBMLPI1280005
ENZOBMLPI1280025
Boc-Gly-Val-Val-CHO
Eiwitten en peptiden
gamma-secretase inhibitor
A novel substrate-based (gamma-40 site) aldehyde inhibitor of gamma-secretase. In whole cells overexpressing amloid ß protein precursor, Boc-GVV-CHO dose-dependently inhibited amyloid ß cleavage. gamma-secretase is an important therapeutic target for strategies designed to lower amyloid ß production.
|