Z-Phe-Ala-FMK
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLPI1380001 BMLPI1380005
ENZOBMLPI1380001EA 130 EUR
ENZOBMLPI1380001 ENZOBMLPI1380005
Z-Phe-Ala-FMK
Eiwitten en peptiden
Caspase inhibitor
Irreversible inhibitor of cathepsins B, L, and S, and likely cathepsin H, as well as cruzain and papain. Has been shown to inhibit effector, but not initator, caspases in vitro, and suppress some forms of apoptosis. Has been used in vivo.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5047e27d7c2b52bf5a46db504c0028d078d066b5&vid=b20286b030c2dc16a85cd9d1c2d58e2defa12af4&prid=web&clid=SAPDAM