Ion channel ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28050100 BML28050500
ENZOBML28050100EA 951 EUR
ENZOBML28050100 ENZOBML28050500
Ion channel ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The Screen-Well® ion channel ligand library contains 71 ion channel blockers and openers for use in characterising and identifying ion channels in individual cells or tissue.