Dopaminergic ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28120100 BML28120500
ENZOBML28120100EA 1330 EUR
ENZOBML28120100 ENZOBML28120500
Dopaminergic ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The Screen-Well® dopaminergic ligands plate contains 80 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The dopaminergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.