Serotonergic ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28130100
BML28130500
ENZOBML28130100EA
2630
EUR
ENZOBML28130100
ENZOBML28130500
Serotonergic ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The Screen-Well® serotonergic ligands plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The serotonergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 680 ligands.