Cholinergic ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28150100 BML28150500
ENZOBML28150100EA 1320 EUR
ENZOBML28150100 ENZOBML28150500
Cholinergic ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The cholinergic ligands plate contains 72 compounds ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The cholinergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.