GABAergic ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28190100 BML28190500
ENZOBML28190100EA 783 EUR
ENZOBML28190100 ENZOBML28190500
GABAergic ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The GABAergic igands library contains 56 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The GABAergics ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 CNS receptor ligands.
Order Now
Specificaties
Environmentally Preferable
Species
Bron