Purinergic ligand library, Screen-Well®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28200100 BML28200500
ENZOBML28200100EA 1850 EUR
ENZOBML28200100 ENZOBML28200500
Purinergic ligand library, Screen-Well®
Celcultuursupplementen en -additieven
The Screen-Well® purinergic and adenosine ligands plate contains 72 CNS receptor ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The purinergic and adenosine ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.
New Product
Order Now
Specificaties
Environmentally Preferable
Species
Bron