Dissolved oxygen probes, TriOxmatic® 700
Leverancier: WTW ONLINE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
201670 201672
662-0508EA 1932 EUR
662-0508 662-0286
Dissolved oxygen probes, TriOxmatic® 700
Elektroden Elektroden voor opgeloste zuurstof
Potentiostatic D.O. sensors with integrated temperature sensor.
  • Integrated lightning protection
  • Sensor monitoring function (SensCheck)
  • Zero current free electrode system
  • Exchangeable membrane head with robust 50 um special membrane
  • Lowest incident flow required
  • Double thermistor system for temperature compensation
  • OxiCal air calibration without accessories
Design according to the minimum requirements defined by ATV (Waste water association) in bulletin M 256 for meters to be used at waste water treatment plants.
To be connected to the EcoLine® orMore Product Information