Measuring cylinders, SUPER DUTY, borosilicate glass, low form, class B, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
213942406 213943608 213944407 213945403
SCOT213942406EA 97.5 EUR
SCOT213942406 SCOT213943608 SCOT213944407 SCOT213945403
Measuring cylinders, SUPER DUTY, borosilicate glass, low form, class B, DURAN®
Cilinders Maatcilinders
DURAN®, borosilicate glass, class B, low form.
  • Very good chemical resistance, high temperature resistance and minimal thermal expansion
  • White easy-to-read scale, highly durable white ceramic
  • Economical
  • Increased safety due to lower breakage risk
  • Supplied with batch certificate
Ideal for demanding applications such as field operations, academic and professional training and frequent stress due to automated washing.
Certificaten: USP Standard; ISO 4788; with printed retrace code (batch identification).