Filter tips, AdvanTip™
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11003-26 68009-13 68009-23
HAMI11003-26EA 181 EUR
HAMI11003-26 HAMI68009-13 HAMI68009-23
Filter tips, AdvanTip™
Pipettips
AdvanTip™ precision pipette tips with hydrophobic PE filter are perfect for routine, high-throughput work. Ideal for PCR, DNA amplification and sequencing.
  • Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
  • Certified free from RNase, DNase, and pyrogens
  • Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
  • Universal fit, can be used with Hamilton SoftGrip™ manual pipettes