Precision thermometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M247302-FLDKD3 M247302-FL
AMARM247302-FLDKD3EA 301 EUR
AMARM247302-FLDKD3 AMARM247302-FL
Precision thermometers
Thermometers Analoge thermometers
These thermometers are suitable for government verification.
  • Round top finish in transparent square plastic case
  • Durable pigment
  • Solid stem and total immersion
Certificaten: Supplied with DIN 12785 certificate.