VWR®, Labelling Tapes, Industrial Grade Polyethylene Tape
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
52B-34WH 52B-3WH 52B-34OR 52B-2YE 52B-12RE 52B-2DB 52B-2TR 52B-2WH 52B-34TR
817-0409EA 22 EUR
817-0409 817-0376 817-0408 817-0406 817-0395 817-0400 817-0404 817-0405 MICR52B-34TR
VWR®, Labelling Tapes, Industrial Grade Polyethylene Tape
Tape
These industrial grade cleanroom tapes are made of PE with acrylic adhesive.
  • Cleanroom support areas
  • Maintenance
  • Clean build projects
For applications where ultra-clean tape is not required. The polyethylene backing with acrylic adhesive has low outgassing and good clean-peel properties. Processed in a controlled environment, tapes are wound on a plastic core and sleeve bagged.
Length: 32,9 m
Different colours available.