Natriumhydroxide 0.5 mol/l (0.5 N) water Volumetric
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Caustic soda
181692.1211 181692.1315
APPC181692.1211EA 30.8 EUR
APPC181692.1211 APPC181692.1315
Natriumhydroxide 0.5 mol/l (0.5 N) water Volumetric
Natriumhydroxide
Absorbs CO₂ from air, miscible with water.
Formule: NaOH
MW: 40 g/mol
Kookpunt: 1390
Smeltpunt: 323
Dichtheid: 1.021 g/cm³
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Specification Test Results
Titer at 20 °C 0.999 - 1.001
Uncertainty See Certificate
Traceability NIST
pH 13.5