Anhydrotetracycline hydrochloride ≥98%, crystalline
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10009542-50 10009542-100 10009542-250
CAYM10009542-50EA 67.4 EUR
CAYM10009542-50 CAYM10009542-100 CAYM10009542-250
Anhydrotetracycline hydrochloride ≥98%, crystalline
Anhydrotetracycline hydrochloride
Anhydrotetracycline (hydrochloride) is a powerful effector in both the tetracycline repressor (TetR) and reverse TetR (revTetR) systems, binding the Tet repressor 35-fold more strongly than Tet.
  • Absorbance: λmax: 224, 272, 428 nm
The tetracycline repressor (TetR) is a transcriptional regulator which normally binds tightly to its palindromic tetO operator DNA, blocking gene expression. Tet causes the repressor to dissociate from the DNA, allowing transcription to occur. A novel reverse TetR (revTetR) requires tetracycline as a co-repressor to bind tetO and block transcription.
Anhydrotetracycline poorly binds the 30S ribosomal subunit, compared to Tet so it does not act as a general inhibitor of translation and is a poor antibiotic. The concentration of anhydrotetracycline that inhibits eukaryotic cell growth is more than a 1,000-fold above the dose that alters transcription through TetR.
Stability: ≥2 years
A crystalline solid
Formule: C₂₂H₂₃ClN₂O₇
MW: 462.883 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00151453
CAS nummer: 13803-65-1