Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Aluminum
43777.GH 43777.HP
43777.GHEA 137 EUR
43777.GH 43777.HP
Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321