Gallium ≥99.9999% (metals basis), pellets, Ø 6 mm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
44324.09 44324.18 44324.30
44324.09EA 127 EUR
44324.09 44324.18 44324.30
Gallium ≥99.9999% (metals basis), pellets, Ø 6 mm
Gallium
Purity: 99.9999% CAS No.: 7440-55-3 Storage Temperature: 2°C to 8°C Physical Form: 0.7 grams/pellet maximum Appearance: Silver grey UN No.: 2803 MDL No.: MFCD00134045
Formule: Ga
Kookpunt: 2403 °C (1013 hPa)
Smeltpunt: 29 °C
Dichtheid: 5,904 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00134045
CAS nummer: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364