Indium 99.99% (trace metals basis), foil, Thickness: 0.127 mm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
318110023 318110092
ACRO318110023EA 130 EUR
ACRO318110023 ACRO318110092
Indium 99.99% (trace metals basis), foil, Thickness: 0.127 mm
Indium
Formule: In
Kookpunt: 2000 °C (760 mmHg)
Smeltpunt: 156 °C
Dichtheid: 7,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971
Order Now
Specification Test Results
Appearance Grey Foil
Total trace metal impurities ≤0.01 %
Specificaties
Environmentally Preferable