Charcoal activated, powder Ash_total_4
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Carbon activated, Activated carbon, Activated charcoal
33302.22 33302.36 33302.A3
33302.22EA 34.8 EUR
33302.22 33302.36 33302.A3
Charcoal activated, powder Ash_total_4
Charcoal activated
CAS No.: 7440-44-0 Appearance: Black UN No.: 1362 MDL No.: MFCD00133992
Formule: C
MW: 12.01 g/mol
Smeltpunt: 3550
Dichtheid: 2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
Bereik: 01-2119488894-16
Merck Index: 13,01818