Graphite electrodes, crater
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14743.KN 40788.KN
14743.KNEA 232 EUR
14743.KN 40788.KN
Graphite electrodes, crater
Elektroden Andere elektroden
Graphite electrodes are used mainly in electric arc furnace steel production.
  • AGKSP grade
  • Ultra F purity
  • Insoluble in water
They are also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings and charging practice.