Tubes, quartz
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75300 75301 75302 75303 75304 75305 75306 75307
TSLA75300EA 25.7 EUR
TSLA75300 TSLA75301 TSLA75302 TSLA75303 TSLA75304 TSLA75305 TSLA75306 TSLA75307
Tubes, quartz
Slangen Transfer Tubing
Fused silica tubes are made from natural quartz to ensure high quality and versatility
  • With an operating range of up to 1200 °C
  • Very good chemical resistance