Serological pipettes, Shorty, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP72115-50510 SP72115-5110
SCERSP72115-50510EA 683 EUR
SCERSP72115-50510 SCERSP72115-5110
Serological pipettes, Shorty, KIMBLE®
Pipetten Serologische pipetten
Shorty pipettes are made from glass and are ideal for use in tissue culture work and other procedures performed under a fume hood or in other confined areas.
  • 'Reverse' and 'negative' graduations increase the versatility of the pipette
  • Calibrated 'to deliver' with blow-out after free flow of liquid has ended
  • Multi-pack bag: One or several pipettes can be removed without contaminating the entire pack
  • Non pyrogenic, sterile, disposable
Verpakking: Individually wrapped in paper/plastic or packed in a multi-pack bag.