Erbium 99,9 Metals basis exTa, rods, Ø 6.35 mm (0.25 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40287.DH 40287.DJ 40287.HF
40287.DHEA 247 EUR
40287.DH 40287.DJ 40287.HF
Erbium 99,9 Metals basis exTa, rods, Ø 6.35 mm (0.25 in)
Erbium
Purity: 99.9% CAS No.: 7440-52-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.29g/mm, Cast rod, diameter is approximate Odour: Odourless
Formule: Er
MW: 167.26 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010987
CAS nummer: 7440-52-0
EINECS: 231-160-1
Order Now
Specificaties
Environmentally Preferable