Terbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Packaged under argon.
12258.03 12258.06 12258.14
12258.03EA 95.6 EUR
12258.03 12258.06 12258.14
Terbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Terbium
Purity: 99.9% CAS No.: 7440-27-9 Physical Form: Packaged under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011256
Formule: Tb
MW: 158.93 g/mol
Kookpunt: 3041
Smeltpunt: 1356
Dichtheid: 8.27 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011256
CAS nummer: 7440-27-9
EINECS: 231-137-6
UN: 3178
ADR: 4.1 II
Merck Index: 12,09300