Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
12430.FF 12430.FI
12430.FFEA 225 EUR
12430.FF 12430.FI
Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Terbium
Purity: 99.9% CAS No.: 7440-27-9 Physical Form: ≈0.52g/25x25mm, Packaged in mylar under argon UN No.: 3178 MDL No.: MFCD00011256
Formule: Tb
MW: 158.93 g/mol
Kookpunt: 3041
Smeltpunt: 1356
Dichtheid: 8.27 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011256
CAS nummer: 7440-27-9
EINECS: 231-137-6
UN: 3178
ADR: 4.1 II
Merck Index: 12,09300