Gold ≥99.999% (metals basis), slug, Premion®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14174.KG 14174.KJ
14174.KGEA 1013 EUR
14174.KG 14174.KJ
Gold ≥99.999% (metals basis), slug, Premion®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Gold
Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈3.35-3.60g/slug, approx. weight - may differ Appearance: Gold Odour: Odourless MDL No.: MFCD00003436
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Dichtheid: 19.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526