Silver hard ≥99.95% (metals basis), foil, hard, Thickness: 0.075 mm (0.003 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00501.FS 00501.GT
00501.FSEA 110 EUR
00501.FS 00501.GT
Silver hard ≥99.95% (metals basis), foil, hard, Thickness: 0.075 mm (0.003 in)
Silver
Purity: 99.95% CAS No.: 7440-22-4 Physical Form: ≈3g/25x150mm, Temper: as rolled Odour: Characteristic MDL No.: MFCD00003397
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Dichtheid: 10.5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647