Silver 99,9985 Metals basis, foil, Premion®, Thickness: 1.0 mm (0.04 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12127.FF 12127.FI 12127.GH
12127.FFEA 83.7 EUR
12127.FF 12127.FI 12127.GH
Silver 99,9985 Metals basis, foil, Premion®, Thickness: 1.0 mm (0.04 in)
Silver
Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈6.6g/25x25mm Odour: Characteristic MDL No.: MFCD00003397
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Dichtheid: 10.5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647