Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14259.BS 14259.BW 14259.VC
14259.BSEA 93.8 EUR
14259.BS 14259.BW 14259.VC
Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈18g/10cm, Temper: as drawn Odour: Characteristic MDL No.: MFCD00003397
Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Smeltpunt: 962 °C
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647