Silver ≥99.97% (metals basis), thin foil, Thickness: 0.005 mm (0.0002 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40748.FF 40748.FI 40748.GH
40748.FFEA 274 EUR
40748.FF 40748.FI 40748.GH
Silver ≥99.97% (metals basis), thin foil, Thickness: 0.005 mm (0.0002 in)
Silver
Purity: 99.97% CAS No.: 7440-22-4 Physical Form: ≈0.03g/25x25mm, Not certified pinhole free Odour: Characteristic MDL No.: MFCD00003397
Microelectronics, contacts and coatings
Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Smeltpunt: 962 °C
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647