Silver ≥99.9% (metals basis), gauze woven from wire 80 mesh, Ø 0.115 mm (0.0045 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40936.FL 40936.GT 40936.HB 40936.FF
40936.FLEA 422 EUR
40936.FL 40936.GT 40936.HB 40936.FF
Silver ≥99.9% (metals basis), gauze woven from wire 80 mesh, Ø 0.115 mm (0.0045 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: Wire Cloth Odour: Characteristic MDL No.: MFCD00003397
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Dichtheid: 10.5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647