Silver ≥99.998% (metals basis), foil, Premion®, annealed, Thickness: 0.1 mm (0.004 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42317.FI 42317.GH 42317.HP
42317.FIEA 73.1 EUR
42317.FI 42317.GH 42317.HP
Silver ≥99.998% (metals basis), foil, Premion®, annealed, Thickness: 0.1 mm (0.004 in)
Silver
Purity: 99.998% CAS No.: 7440-22-4 Physical Form: ≈2.6g/50x50mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397
Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Smeltpunt: 962 °C
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647