Scandium ≥99.6% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43499.HF 43499.DH
43499.HFEA 845 EUR
43499.HF 43499.DH
Scandium ≥99.6% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Scandium
Purity: 99.6% CAS No.: 7440-20-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.53g/mm, Cast rod, diameter is approximate Appearance: Grey Odour: Odourless MDL No.: MFCD00016323
Formule: Sc
MW: 44.96 g/mol
Kookpunt: 2730
Smeltpunt: 1541
Dichtheid: 2.989 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016323
CAS nummer: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537