Heat resistant welding gloves, ActivArmr® 43-217
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43-217/10 43-217/11 43-217/9
ANSE43-217/10EA 94.7 EUR
ANSE43-217/10 ANSE43-217/11 ANSE43-217/9
Heat resistant welding gloves, ActivArmr® 43-217
Handschoenen
This TIG welding glove made from full grain goatskin provides EN 12477 Type-B protection for precise welding and thermal tasks.
  • Provides protection from heat, flames, sparks and weld spatter
  • 3-dimensional thumb design greatly increases fine dexterity for precise welding control
  • Premium full grain goatskin and welted seams with para-aramid thread ensure durability in use
  • Long split cowhide cuff extends protection to the forearms
Suitable for TIG welding, as well as grinding, brazing, metal fabrication and maintenance.
Certificaten: EN 407 (41XX4X), EN 12477 Type-B; Cat. III