Neck tube, FR19
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR19BKR FR19NAR
PRTWFR19BKREA 25.4 EUR
PRTWFR19BKR PRTWFR19NAR
Neck tube, FR19
Werkkleding
This anti-static neck tube is flame resistant so can be worn when dealing with flame or heat. Made from Modaflame knit (60% Modacrylic, 39% cotton and 1% carbon fibre).
  • Ideal for protection in multiple work places
  • Extremely comfortable
  • Provides protection for the entire neck area
Weight: 200 g/m²