Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.127 mm (0.005 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00236.FF 00236.FQ 00236.FY 00236.FI
00236.FFEA 46.4 EUR
00236.FF 00236.FQ 00236.FY 00236.FI
Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.127 mm (0.005 in)
Niobium
Purity: 99.97% CAS No.: 7440-03-1 Physical Form: ≈0.68g/25x25mm Appearance: Grey or silver Odour: Odourless MDL No.: MFCD00011126
Formule: Nb
Kookpunt: 3700 °C (1013 hPa)
Smeltpunt: 2468 °C
Dichtheid: 8,57 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011126
CAS nummer: 7440-03-1
EINECS: 231-113-5