Iridium 99.9% (trace metals basis), powder
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
198095000 198090100
ACRO198095000EA 408 EUR
ACRO198095000 ACRO198090100
Iridium 99.9% (trace metals basis), powder
Iridium
Formule: Ir
MW: 192.22 g/mol
Kookpunt: 5300
Smeltpunt: 2450 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011062
CAS nummer: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1 II
Order Now
Specification Test Results
Appearance Grey Powder
Total trace metal impurities ≤0.1 %